Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Glycerol solution
Synonym(s):
1,2,3-Propanetriol
Linear Formula:
HOCH
2
CH(OH)CH
2
OH · aq
CAS Number:
56-81-5
Molecular Weight:
92.09 (anhydrous basis)
EC Number:
200-289-5
Products
Product Category
glycerols (1)
lipids (1)
Brand
Sigma-Aldrich (2)
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Special Grade
Puriss (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
49781
Glycerol solution,
83.5-89.5% (T)
Expand
View Pricing
15524
Glycerol solution,
puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Expand
View Pricing