Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Glycerol-1,1,2,3,3-d
5
Linear Formula:
(HOCD
2
)
2
CDOH
CAS Number:
62502-71-0
Molecular Weight:
97.12
Products
Product Category
alcohols (3)
glycerols (3)
stable isotopes (3)
polyols (1)
Brand
Sigma-Aldrich (2)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Special Grade
GMP (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
454524
Glycerol-1,1,2,3,3-d
5
,
98 atom % D
Expand
View Pricing
W6926
Glycerol-1,1,2,3,3-d
5
,
API for Clinical Studies, (API for Clinical Studies), ≥98 atom % D, ≥98% (CP)
Expand
View Pricing
661473
Glycerol-1,1,2,3,3-d
5
,
endotoxin tested, 98 atom % D
Expand
View Pricing