
Fusidic acid

Synonym(s):
Fusidic acid
Empirical Formula (Hill Notation):
C31H48O6
CAS Number:
Molecular Weight:
516.71
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3