Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Fritted Purge Sampler
Products
Brand
Supelco (3)
Manufacturer
Agency
Compatibility
Quality Segment
Application
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
22781
Fritted Purge Sampler,
size 5 mL, Tekmar
®
14-2337-024
Expand
View Pricing
22789
Fritted Purge Sampler,
size 25 mL, Tekmar
®
14-2334-024
Expand
View Pricing
22749
Fritted Purge Sampler,
size 25 mL, top stem, Tekmar
®
14-4007-024
Expand
View Pricing