Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Formic acid 98% - 100%
Synonym(s):
Formic acid
Linear Formula:
HCOOH
CAS Number:
64-18-6
Molecular Weight:
46.03
Products
Product Category
buffers (2)
Brand
Supelco (4)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Agency
Quality Segment
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
5.33002
Formic acid 98% - 100%,
for LC-MS LiChropur
™
Expand
View Pricing
1.00264
Formic acid 98% - 100%,
for analysis EMSURE
®
ACS,Reag. Ph Eur
Expand
View Pricing
5.43804
Formic acid 98% - 100%,
for HPLC LiChropur
™
Expand
View Pricing
1.11670
Formic acid 98% - 100%,
Suprapur
®
Expand
View Pricing