Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Formaldehyde-2,4-DNPH Solution
Linear Formula:
H
2
C=NNHC
6
H
3
(NO
2
)
2
CAS Number:
1081-15-8
Molecular Weight:
210.15
EC Number:
200-835-2
Products
Brand
Supelco (1)
Formula Weight
Product Line
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
certified reference material (1)
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
CRM4M7177
Formaldehyde-2,4-DNPH Solution,
certified reference material,
Trace
CERT
®
, 100 μg/mL in acetonitrile (as aldehyde), ampule of 5 × 1 mL
Expand
View Pricing