
Fmoc-Lys(Me,Boc)-OH

Empirical Formula (Hill Notation):
C27H34N2O6
Molecular Weight:
482.57
Products
Brand
Formula Weight
Purity
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing