Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Fluorouracil
Synonym(s):
5-Fluorouracil, 2,4-Dihydroxy-5-fluoropyrimidine, 5-FU, 5-Fluoro-2,4(1
H
,3
H
)-pyrimidinedione
Empirical Formula (Hill Notation):
C
4
H
3
FN
2
O
2
CAS Number:
51-21-8
Molecular Weight:
130.08
EC Number:
200-085-6
Products
Brand
Sigma-Aldrich (1)
Supelco (1)
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
pharmaceutical primary standard (3)
certified reference material (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
1279000
Fluorouracil,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
F8423
Fluorouracil,
meets USP testing specifications
Expand
View Pricing
PHR1227
Fluorouracil,
Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing
F0250000
Fluorouracil,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing
BP995
Fluorouracil,
British Pharmacopoeia (BP) Reference Standard
Expand
View Pricing