Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Fluorine doped tin oxide coated glass slide
Synonym(s):
FTO glass
EC Number:
242-159-0
Products
Brand
Sigma-Aldrich (11)
Chemical Composition
Diameter (mm)
Length (mm)
Package Size
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
735167
Fluorine doped tin oxide coated glass slide,
L × W × thickness 300 mm × 300 mm × 2.2 mm, surface resistivity ~7 Ω/sq
Expand
View Pricing
735140
Fluorine doped tin oxide coated glass slide,
L × W × thickness 50 mm × 50 mm × 2.2 mm, surface resistivity ~7 Ω/sq
Expand
View Pricing
735159
Fluorine doped tin oxide coated glass slide,
L × W × thickness 100 mm × 100 mm × 2.3 mm, surface resistivity ~7 Ω/sq
Expand
View Pricing
735256
Fluorine doped tin oxide coated glass slide,
L × W × D 100 mm × 100 mm × 2.2 mm, surface resistivity ~13 Ω/sq
Expand
View Pricing
735248
Fluorine doped tin oxide coated glass slide,
L × W × D 50 mm × 50 mm × 2.2 mm, surface resistivity ~13 Ω/sq
Expand
View Pricing
735175
Fluorine doped tin oxide coated glass slide,
L × W × thickness 50 mm × 50 mm × 3 mm, surface resistivity ~8 Ω/sq
Expand
View Pricing
735264
Fluorine doped tin oxide coated glass slide,
L × W × D 300 mm × 300 mm × 2.3 mm, surface resistivity ~13 Ω/sq
Expand
View Pricing
735183
Fluorine doped tin oxide coated glass slide,
surface resistivity ~8 Ω/sq
Expand
View Pricing
735205
Fluorine doped tin oxide coated glass slide,
L × W × D 50 mm × 50 mm × 3 mm, surface resistivity ~10 Ω/sq
Expand
View Pricing
735213
Fluorine doped tin oxide coated glass slide,
L × W × D 100 mm × 100 mm × 3 mm, surface resistivity ~10 Ω/sq
Expand
View Pricing
735221
Fluorine doped tin oxide coated glass slide,
L × W × D 300 mm × 300 mm × 3 mm, surface resistivity ~10 Ω/sq
Expand
View Pricing
735191
Fluorine doped tin oxide coated glass slide,
L × W × thickness 300 mm × 300 mm × 3.2 mm, surface resistivity ~8 Ω/sq
Expand
View Pricing