
Fipronil sulfone

Synonym(s):
5-Amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-[(trifluoromethyl)sulfonyl]-1H-pyrazole-3-carbonitrile
Empirical Formula (Hill Notation):
C12H4Cl2F6N4O2S
CAS Number:
Molecular Weight:
453.15
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2