Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Finntip
®
Flex pipette tips
Products
Product Category
pipette tips (12)
Manufacturer
Volume Range (µL)
Color
Package Size
Sterility
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
Z677965
Finntip
®
Flex pipette tips,
Finntip Flex 300, non-sterile
Expand
View Pricing
Z677833
Finntip
®
Flex pipette tips,
Finntip Flex 10, sterile
Expand
View Pricing
Z677922
Finntip
®
Flex pipette tips,
Finntip Flex 1000, sterile
Expand
View Pricing
Z677914
Finntip
®
Flex pipette tips,
Finntip Flex 1000, non-sterile
Expand
View Pricing
Z677825
Finntip
®
Flex pipette tips,
Finntip Flex 10, non-sterile, pack of 1000 ea (Bag)
Expand
View Pricing
Z677868
Finntip
®
Flex pipette tips,
Finntip Flex 200, non-sterile
Expand
View Pricing
Z677876
Finntip
®
Flex pipette tips,
Finntip Flex 200, sterile
Expand
View Pricing
Z677884
Finntip
®
Flex pipette tips,
Finntip Flex 200, non-sterile
Expand
View Pricing
Z677930
Finntip
®
Flex pipette tips,
Finntip Flex 1000, non-sterile
Expand
View Pricing
Z677841
Finntip
®
Flex pipette tips,
Finntip Flex 10, non-sterile
Expand
View Pricing
Z677957
Finntip
®
Flex pipette tips,
Finntip Flex 300, non-sterile
Expand
View Pricing
Z677949
Finntip
®
Flex pipette tips,
Finntip Flex 300, non-sterile
Expand
View Pricing