Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Finnpipette
®
Stepper pipette tips
Products
Product Category
pipette tips (8)
Manufacturer
Color
Package Size
Sterility
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
Z369187
Finnpipette
®
Stepper pipette tips,
maximum volume 5 mL, sterile
Expand
View Pricing
Z369152
Finnpipette
®
Stepper pipette tips,
maximum volume 0.5 mL, sterile
Expand
View Pricing
Z369144
Finnpipette
®
Stepper pipette tips,
maximum volume 50 mL
Expand
View Pricing
Z369101
Finnpipette
®
Stepper pipette tips,
maximum volume 5 mL
Expand
View Pricing
Z369179
Finnpipette
®
Stepper pipette tips,
maximum volume 2.5 mL, sterile
Expand
View Pricing
Z369160
Finnpipette
®
Stepper pipette tips,
maximum volume 1.25 mL, sterile
Expand
View Pricing
Z369063
Finnpipette
®
Stepper pipette tips,
maximum volume 0.5 mL
Expand
View Pricing
Z369071
Finnpipette
®
Stepper pipette tips,
maximum volume 1.25 mL
Expand
View Pricing