Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Fecralloy - Iron-Chromium, Fe72.8Cr22Al5Y0.1Zr0.1
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Length (mm)
Filter Products
Showing 1-20 of 134
Page 1 of 7
Page 1 of 7
Compare
Product Number
Description
Pricing
GF98356078
Fecralloy - Iron-Chromium, Fe72.8Cr22Al5Y0.1Zr0.1,
500mm tube, 6.0mm outside diameter, 0.4mm wall thickness, 5.2mm inside diameter
Expand
View Pricing
GF11284679
Fecralloy - Iron-Chromium, Fe72.8Cr22Al5Y0.1Zr0.1,
foil, 1m coil, 20mm coil width, 0.1mm thickness, as rolled
Expand
View Pricing
GF05983139
Fecralloy - Iron-Chromium, Fe72.8Cr22Al5Y0.1Zr0.1,
500mm tube, 15mm outside diameter, 1.5mm wall thickness, 12mm inside diameter
Expand
View Pricing
GF88759681
Fecralloy - Iron-Chromium, Fe72.8Cr22Al5Y0.1Zr0.1,
100mm tube, 12.7mm outside diameter, 0.8mm wall thickness, 11.1mm inside diameter
Expand
View Pricing
GF51383864
Fecralloy - Iron-Chromium, Fe72.8Cr22Al5Y0.1Zr0.1,
100m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF52190827
Fecralloy - Iron-Chromium, Fe72.8Cr22Al5Y0.1Zr0.1,
1000mm rod, 1.8mm diameter
Expand
View Pricing
GF12698556
Fecralloy - Iron-Chromium, Fe72.8Cr22Al5Y0.1Zr0.1,
foil, 70 x 70mm, 0.2mm thickness, annealed
Expand
View Pricing
GF07369588
Fecralloy - Iron-Chromium, Fe72.8Cr22Al5Y0.1Zr0.1,
foil, 180 x 180mm, 0.05mm thickness, annealed, light tight (lt)
Expand
View Pricing
GF21822308
Fecralloy - Iron-Chromium, Fe72.8Cr22Al5Y0.1Zr0.1,
foil, 40 x 40mm, 0.01mm thickness, as rolled, not light tested (nlt)
Expand
View Pricing
GF26977876
Fecralloy - Iron-Chromium, Fe72.8Cr22Al5Y0.1Zr0.1,
sheet, 100 x 100mm, 3mm thickness, as rolled
Expand
View Pricing
GF26361817
Fecralloy - Iron-Chromium, Fe72.8Cr22Al5Y0.1Zr0.1,
foil, 90 x 90mm, 0.05mm thickness, annealed, light tight (lt)
Expand
View Pricing
GF12191330
Fecralloy - Iron-Chromium, Fe72.8Cr22Al5Y0.1Zr0.1,
1000mm rod, 3.2mm diameter
Expand
View Pricing
GF21919990
Fecralloy - Iron-Chromium, Fe72.8Cr22Al5Y0.1Zr0.1,
200mm rod, 1.8mm diameter
Expand
View Pricing
GF62483328
Fecralloy - Iron-Chromium, Fe72.8Cr22Al5Y0.1Zr0.1,
100mm rod, 1.8mm diameter
Expand
View Pricing
GF85149142
Fecralloy - Iron-Chromium, Fe72.8Cr22Al5Y0.1Zr0.1,
foil, 50 x 50mm, 2.0mm thickness, as rolled
Expand
View Pricing
GF57647166
Fecralloy - Iron-Chromium, Fe72.8Cr22Al5Y0.1Zr0.1,
foil, 200 x 1000mm, 0.5mm thickness, as rolled
Expand
View Pricing
GF72124499
Fecralloy - Iron-Chromium, Fe72.8Cr22Al5Y0.1Zr0.1,
foil, 50 x 100mm, 0.3mm thickness, annealed
Expand
View Pricing
GF07853845
Fecralloy - Iron-Chromium, Fe72.8Cr22Al5Y0.1Zr0.1,
foil, 100 x 100mm, 2.0mm thickness, as rolled
Expand
View Pricing
GF37008086
Fecralloy - Iron-Chromium, Fe72.8Cr22Al5Y0.1Zr0.1,
foil, 200 x 500mm, 0.5mm thickness, as rolled
Expand
View Pricing
GF48185758
Fecralloy - Iron-Chromium, Fe72.8Cr22Al5Y0.1Zr0.1,
foil, 100 x 100mm, 0.5mm thickness, as rolled
Expand
View Pricing
Page 1 of 7
Page 1 of 7