Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
EX-CELL
®
NS0 Serum-Free Medium for NS0 Cells
Products
Product Category
media (2)
Brand
SAFC (2)
Physical Form
HEPES (mM)
L-Glutamine
Quality Segment
Sterilization Method
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
14650C
EX-CELL
®
NS0 Serum-Free Medium for NS0 Cells,
Animal-component free, Protein-free, Chemically defined, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Expand
View Pricing
24651C
EX-CELL
®
NS0 Serum-Free Medium for NS0 Cells,
Animal-component free, Protein-free, Chemically defined, dry powder, without L-glutamine, without sodium bicarbonate, suitable for cell culture
Expand
View Pricing