Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Exact-Temp Coated bottle thermometer
Products
Color
Length (mm)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
Z742061
Exact-Temp Coated bottle thermometer,
for oven, temperature +20 to +130 °C, precision: ±1 °C
Expand
View Pricing
Z742052
Exact-Temp Coated bottle thermometer,
for Freezer, temperature -30-0 °C, precision: ±0.5 °C
Expand
View Pricing
Z742060
Exact-Temp Coated bottle thermometer,
for incubator, temperature +25 to +45 °C, precision: ±0.1 °C
Expand
View Pricing
Z742051
Exact-Temp Coated bottle thermometer,
for Freezer, temperature -25 to -5 °C, precision: ±0.1 °C
Expand
View Pricing
Z742050
Exact-Temp Coated bottle thermometer,
for ultralow Freezers, temperature -90 to +20 °C, precision: ±1 °C
Expand
View Pricing
Z742057
Exact-Temp Coated bottle thermometer,
for incubator, temperature +15 to +30 °C, precision: ±0.1 °C
Expand
View Pricing
Z742059
Exact-Temp Coated bottle thermometer,
for incubator, temperature +18 to +50 °C, precision: ±0.5 °C
Expand
View Pricing