Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Exact-Temp bottle thermometer
Products
Color
Length (mm)
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
Z741985
Exact-Temp bottle thermometer,
for ultralow Freezers, temperature -90 to +20 °C, accuracy: 1 °C
Expand
View Pricing
Z741988
Exact-Temp bottle thermometer,
for refrigerator, temperature -2 to +10 °C, accuracy: 0.1 °C
Expand
View Pricing
Z741996
Exact-Temp bottle thermometer,
for oven, temperature +35 to +200 °C, accuracy: 1 °C
Expand
View Pricing
Z741987
Exact-Temp bottle thermometer,
for Freezers, temperature -30-0 °C, accuracy: 0.1 °C
Expand
View Pricing
Z741995
Exact-Temp bottle thermometer,
for oven, temperature +20 to +130 °C, accuracy: 1 °C
Expand
View Pricing
Z741992
Exact-Temp bottle thermometer,
for incubator, temperature +15 to +30 °C, accuracy: 0.1 °C
Expand
View Pricing
Z741993
Exact-Temp bottle thermometer,
for incubator, temperature +18 to +50 °C, accuracy: 0.5 °C
Expand
View Pricing
Z741994
Exact-Temp bottle thermometer,
for incubator, temperature +25 to +45 °C, accuracy: 0.1 °C
Expand
View Pricing
Z741989
Exact-Temp bottle thermometer,
for refrigerator, temperature -5 to +15 °C, accuracy: 0.1 °C
Expand
View Pricing
Z741997
Exact-Temp bottle thermometer,
for oven, temperature +50 to +115 °C, accuracy: 0.5 °C
Expand
View Pricing
Z741986
Exact-Temp bottle thermometer,
for Freezers, temperature -25 to -5 °C, accuracy: 0.1 °C
Expand
View Pricing