![Ethylenediaminetetraacetic acid iron(III) sodium salt hydrate 12-14% Fe basis](/deepweb/assets/sigmaaldrich/product/structures/277/671/b8264704-a8ee-4ecd-843d-5541d350d548/640/b8264704-a8ee-4ecd-843d-5541d350d548.png)
Ethylenediaminetetraacetic acid iron(III) sodium salt hydrate
![Ethylenediaminetetraacetic acid iron(III) sodium salt hydrate 12-14% Fe basis](/deepweb/assets/sigmaaldrich/product/structures/277/671/b8264704-a8ee-4ecd-843d-5541d350d548/640/b8264704-a8ee-4ecd-843d-5541d350d548.png)
Synonym(s):
Iron(III)-sodium ethylenediaminetetraacetate hydrate
Linear Formula:
(OOCCH2)2NCH2CH2N(CH2COO)2FeNa · xH2O
CAS Number:
Molecular Weight:
367.05
Products
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing