
Estradiol Related Compound C

Synonym(s):
β-Estradiol-6-one, 1,3,5-Estratriene-3,17β-diol-6-one, 3,17β-Dihydroxy-1,3,5(10)-estratrien-6-one, 6-Ketoestradiol
Empirical Formula (Hill Notation):
C18H22O3
CAS Number:
Molecular Weight:
286.37
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2