Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf twin.tec
®
PCR Plates 96
Products
Product Category
plates (10)
Color
Manufacturer
Binding Type
Wells
Color
Package Size
Sterility
Technique
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
EP0030129512
Eppendorf twin.tec
®
PCR Plates 96,
Skirted, LoBind, PCR clean, working volume 150 μL, colorless, pack of 25
Expand
View Pricing
EP0030129504
Eppendorf twin.tec
®
PCR Plates 96,
Semi-Skirted, LoBind, PCR clean, working volume 250 μL, colorless, pack of 25
Expand
View Pricing
EP0030129580
Eppendorf twin.tec
®
PCR Plates 96,
PCR clean, skirted, blue, pkg of 25 ea
Expand
View Pricing
EP0030133307
Eppendorf twin.tec
®
PCR Plates 96,
Unskirted, PCR clean, volume 150 μL, colorless, pack of 20
Expand
View Pricing
EP0030133366
Eppendorf twin.tec
®
PCR Plates 96,
clear plate, unskirted, conical bottom colorless wells, capacity 250 μL, colorless, pkg of 20 ea
Expand
View Pricing
EP0030129660
Eppendorf twin.tec
®
PCR Plates 96,
PCR clean, skirt, green, pkg of 25 ea
Expand
View Pricing
EP0030133374
Eppendorf twin.tec
®
PCR Plates 96,
clear plate, divisible, unskirted, conical bottom colorless wells, capacity 250 μL, colorless, unskirted, divisible, pkg of 20 ea
Expand
View Pricing
EP0030129679
Eppendorf twin.tec
®
PCR Plates 96,
PCR clean, skirted, yellow, pkg of 25 ea
Expand
View Pricing
EP0030133358
Eppendorf twin.tec
®
PCR Plates 96,
divisible, unskirted, clear plate, conical bottom colorless wells, colorless, unskirted, low profile, divisible, pkg of 20 ea
Expand
View Pricing
EP0030129610
Eppendorf twin.tec
®
PCR Plates 96,
Semi-Skirted, Forensic DNA Grade, working volume 250 μL, colorless, pack of 10 (individually wrapped)
Expand
View Pricing
EP0030129598
Eppendorf twin.tec
®
PCR Plates 96,
PCR clean, skirted, red, pkg of 25 ea
Expand
View Pricing
EP0030129601
Eppendorf twin.tec
®
PCR Plates 96,
Skirted, Forensic DNA Grade, working volume 150 μL, colorless, pack of 10 (individually wrapped)
Expand
View Pricing
EP0030133404
Eppendorf twin.tec
®
PCR Plates 96,
divisible, unskirted, blue piston, conical bottom colorless wells, blue, capacity 250 μL, unskirted, divisible, pkg of 20 ea
Expand
View Pricing
EP0030133382
Eppendorf twin.tec
®
PCR Plates 96,
divisible, unskirted, blue plate, conical bottom colorless wells, blue, unskirted, low profile, divisible, pkg of 20 ea
Expand
View Pricing
EP0030113560
Eppendorf twin.tec
®
PCR Plates 96,
skirt, PCR clean, working volume 150 μL, colorless (with 2D SafeCode), pkg of 25 ea
Expand
View Pricing
EP0030133331
Eppendorf twin.tec
®
PCR Plates 96,
blue plate, low profile, unskirted, conical bottom colorless wells, unskirted, low profile, blue, pkg of 20 ea
Expand
View Pricing