Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Serological Pipets, Sterile Grade
Products
Product Category
pipettes (6)
Manufacturer
Capacity (mL)
Color
Package Size
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
EP0030127730
Eppendorf
®
Serological Pipets, Sterile Grade,
red, capacity 25 mL, pack of 200 ea (4 bags × 50 pcs.)
Expand
View Pricing
EP0030127722
Eppendorf
®
Serological Pipets, Sterile Grade,
orange, capacity 10 mL, pack of 400 ea (4 bags × 100 pcs.)
Expand
View Pricing
EP0030127749
Eppendorf
®
Serological Pipets, Sterile Grade,
violet, capacity 50 mL, pack of 160 ea (4 bags × 40 pcs.)
Expand
View Pricing
EP0030127714
Eppendorf
®
Serological Pipets, Sterile Grade,
blue, capacity 5 mL, pack of 400 ea (4 bags × 100 pcs.)
Expand
View Pricing
EP0030127706
Eppendorf
®
Serological Pipets, Sterile Grade,
green, capacity 2 mL, pack of 600 (4 bags × 150 pcs.)
Expand
View Pricing
EP0030127692
Eppendorf
®
Serological Pipets, Sterile Grade,
yellow, capacity 1 mL, pack of 800 (4 bags × 200 pcs.)
Expand
View Pricing