Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf Research
®
plus Pipette, fixed volume
Products
Product Category
pipettors (11)
Manufacturer
Volume (µL)
Color
Package Size
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
EP3124000121
Eppendorf Research
®
plus Pipette, fixed volume,
single-channel, volume 1000 μL, blue
Expand
View Pricing
EP3124000083
Eppendorf Research
®
plus Pipette, fixed volume,
single-channel, volume 200 μL, yellow
Expand
View Pricing
EP3124000024
Eppendorf Research
®
plus Pipette, fixed volume,
single-channel, volume 10 μL, yellow
Expand
View Pricing
EP3124000075
Eppendorf Research
®
plus Pipette, fixed volume,
single-channel, volume 100 μL, yellow
Expand
View Pricing
EP3124000113
Eppendorf Research
®
plus Pipette, fixed volume,
single-channel, volume 500 μL, blue
Expand
View Pricing
EP3124000040
Eppendorf Research
®
plus Pipette, fixed volume,
single-channel, volume 20 μL, yellow
Expand
View Pricing
EP3124000105
Eppendorf Research
®
plus Pipette, fixed volume,
single-channel, volume 250 μL, blue
Expand
View Pricing
EP3124000059
Eppendorf Research
®
plus Pipette, fixed volume,
single-channel, volume 25 μL, yellow
Expand
View Pricing
EP3124000032
Eppendorf Research
®
plus Pipette, fixed volume,
single-channel, volume 20 μL, gray
Expand
View Pricing
EP3124000067
Eppendorf Research
®
plus Pipette, fixed volume,
single-channel, volume 50 μL, yellow
Expand
View Pricing
EP3124000091
Eppendorf Research
®
plus Pipette, fixed volume,
single-channel, volume 200 μL, blue
Expand
View Pricing