Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf Research
®
Plus G
Products
Product Category
pipettors (25)
Color
Manufacturer
Volume (ml)
Volume (µL)
Package Size
Filter Products
Showing 1-20 of 26
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
EP3123000918
Eppendorf Research
®
Plus G,
3-pack, option 2, 2-20 uL/20-200 uL/100-1000 uL, includes epT.I.P.S.
®
box
Expand
View Pricing
EP3123000063
Eppendorf Research
®
Plus G,
single-channel, variable, volume (100—1,000 μL), blue, includes epT.I.P.S.
®
box
Expand
View Pricing
EP3123000900
Eppendorf Research
®
Plus G,
3-pack, Option 1, 0.5-10 uL/10-100 uL/100-1000 uL, includes epT.I.P.S.
®
box
Expand
View Pricing
EP3123000012
Eppendorf Research
®
Plus G,
single-channel, variable, volume (0.1-2.5 μL), dark gray, includes epT.I.P.S.
®
box
Expand
View Pricing
EP3123000020
Eppendorf Research
®
Plus G,
single-channel, variable 0.5 – 10 uL, medium gray includes epT.I.P.S.(R) box, volume (0.5-10 μL), medium gray, includes epT.I.P.S.
®
box
Expand
View Pricing
EP3123000055
Eppendorf Research
®
Plus G,
single-channel, variable, volume (20-200 μL), yellow, includes epT.I.P.S.
®
box
Expand
View Pricing
EP3125000060
Eppendorf Research
®
Plus G,
12-channel, variable, volume (30-300 μL), orange
Expand
View Pricing
EP3123000071
Eppendorf Research
®
Plus G,
single-channel, variable, volume (0.5-5 mL), purple, includes epT.I.P.S.
®
sample BAG
Expand
View Pricing
EP3125000044
Eppendorf Research
®
Plus G,
12-channel, variable, volume (10-100 μL), yellow
Expand
View Pricing
EP3123000080
Eppendorf Research
®
Plus G,
single-channel, variable, volume (1-10 mL), turquoise, includes epT.I.P.S.
®
SAMPLE BAG
Expand
View Pricing
EP3125000052
Eppendorf Research
®
Plus G,
8-channel, variable, volume (30-300 μL), orange
Expand
View Pricing
EP3123000047
Eppendorf Research
®
Plus G,
single-channel, variable, volume (10-100 μL), yellow, includes epT.I.P.S.
®
box
Expand
View Pricing
EP3125000036
Eppendorf Research
®
Plus G,
8-channel, variable, volume (10-100 μL), yellow
Expand
View Pricing
EP3123000039
Eppendorf Research
®
Plus G,
single-channel, variable, volume (2-20 μL), yellow, includes epT.I.P.S.
®
box
Expand
View Pricing
EP3123000926
Eppendorf Research
®
Plus G,
3-pack, option 3, 100-1000 uL/0,5-5 mL/1-10 mL, includes epT.I.P.S.
®
box
Expand
View Pricing
EP3123000098
Eppendorf Research
®
Plus G,
single-channel, variable, volume (2-20 μL), light gray, includes epT.I.P.S.
®
box
Expand
View Pricing
EP3125000010
Eppendorf Research
®
Plus G,
8-channel, variable, volume (0.5-10 μL), medium gray
Expand
View Pricing
EP3125000214
Eppendorf Research
®
Plus G,
8-channel, tip volume 120-1200 μL
Expand
View Pricing
EP3123000101
Eppendorf Research
®
Plus G,
Esingle-channel, variable, volume (30-300 μL), orange, includes epT.I.P.S.
®
box
Expand
View Pricing
EP3125000028
Eppendorf Research
®
Plus G,
12-channel, variable, volume (0.5-10 μL), medium gray
Expand
View Pricing
Page 1 of 2
Page 1 of 2