Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Reference
®
2 Variable Volume Pipettor
Products
Product Category
pipettors (6)
Manufacturer
Volume (µL)
Color
Package Size
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
EP4926000034
Eppendorf
®
Reference
®
2 Variable Volume Pipettor,
8-channel, incl. epT.I.P.S.
®
Box, volume 10-100 μL, yellow
Expand
View Pricing
EP4926000018
Eppendorf
®
Reference
®
2 Variable Volume Pipettor,
8-channel, incl. epT.I.P.S.
®
Box, volume 0.5-10 μL, gray
Expand
View Pricing
EP4926000050
Eppendorf
®
Reference
®
2 Variable Volume Pipettor,
8-channel, incl. epT.I.P.S.
®
Box, volume 30-300 μL, orange
Expand
View Pricing
EP4926000069
Eppendorf
®
Reference
®
2 Variable Volume Pipettor,
12-channel, incl. epT.I.P.S.
®
Box, volume 30-300 μL, orange
Expand
View Pricing
EP4926000026
Eppendorf
®
Reference
®
2 Variable Volume Pipettor,
12-channel, incl. epT.I.P.S.
®
Box, volume 0.5-10 μL, gray
Expand
View Pricing
EP4926000042
Eppendorf
®
Reference
®
2 Variable Volume Pipettor,
12-channel, incl. epT.I.P.S.
®
Box, volume 10-100 μL, yellow
Expand
View Pricing