Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
ep Dualfilter T.I.P.S.
®
SealMax
Products
Product Category
pipette tips (6)
Color
Volume (µL)
Package Size
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
EP0030078691
Eppendorf
®
ep Dualfilter T.I.P.S.
®
SealMax,
PCR clean, sterile, tip volume × L 0.1-10 μL × 34 mm, graydark, case of 960 tips (10 racks x 96 tips)
Expand
View Pricing
EP0030078730
Eppendorf
®
ep Dualfilter T.I.P.S.
®
SealMax,
PCR clean, sterile, tip volume × L 20-300 μL × 55 mm, orange, case of 960 tips (10 racks x 96 tips)
Expand
View Pricing
EP0030078721
Eppendorf
®
ep Dualfilter T.I.P.S.
®
SealMax,
PCR clean, sterile, tip volume × L 2-200 μL × 55 mm, yellow, case of 960 tips (10 racks x 96 tips)
Expand
View Pricing
EP0030078748
Eppendorf
®
ep Dualfilter T.I.P.S.
®
SealMax,
PCR clean, sterile, tip volume × L 50-1000 μL × 76 mm, blue, case of 960 tips (10 racks x 96 tips)
Expand
View Pricing
EP0030078713
Eppendorf
®
ep Dualfilter T.I.P.S.
®
SealMax,
PCR clean, sterile, tip volume × L 2-100 μL × 53 mm, yellow, case of 960 tips (10 racks x 96 tips)
Expand
View Pricing
EP0030078705
Eppendorf
®
ep Dualfilter T.I.P.S.
®
SealMax,
PCR clean, sterile, tip volume × L 0.5-20 μL × 46 mm, light gray, case of 960 tips (10 racks x 96 tips)
Expand
View Pricing