Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Empore
™
Extraction Disk solid phase extraction (SPE) Cartridge
Products
Brand
Supelco (3)
Manufacturer
ID (mm)
Volume (ml)
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
66873-U
Empore
™
Extraction Disk solid phase extraction (SPE) Cartridge,
matrix active group C18-SD, volume 6 mL, pk of 30
Expand
View Pricing
66872-U
Empore
™
Extraction Disk solid phase extraction (SPE) Cartridge,
matrix active group C18-SD, volume 3 mL, pk of 50
Expand
View Pricing
66871-U
Empore
™
Extraction Disk solid phase extraction (SPE) Cartridge,
matrix active group C18-SD, volume 1 mL, pk of 100
Expand
View Pricing