Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Duran
®
Youtility bottle, GL 45, complete
Synonym(s):
Duran
®
Youtility
Products
Product Category
bottles (8)
Capacity (mL)
Cap
Color
Package Size
Sterility
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
Z742010
Duran
®
Youtility bottle, GL 45, complete,
clear glass, capacity 1000 mL
Expand
View Pricing
Z742009
Duran
®
Youtility bottle, GL 45, complete,
clear glass, capacity 500 mL
Expand
View Pricing
Z742008
Duran
®
Youtility bottle, GL 45, complete,
clear glass, capacity 250 mL
Expand
View Pricing
Z742014
Duran
®
Youtility bottle, GL 45, complete,
amber glass, capacity 1000 mL
Expand
View Pricing
Z742006
Duran
®
Youtility bottle, GL 45, complete,
clear glass, capacity 125 mL
Expand
View Pricing
Z742012
Duran
®
Youtility bottle, GL 45, complete,
amber glass, capacity 250 mL
Expand
View Pricing
Z742013
Duran
®
Youtility bottle, GL 45, complete,
amber glass, capacity 500 mL
Expand
View Pricing
Z742011
Duran
®
Youtility bottle, GL 45, complete,
amber glass, capacity 125 mL
Expand
View Pricing