Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Duran
®
Laboratory Bottle
Products
Product Category
bottles (8)
Manufacturer
Capacity (mL)
Color
Vial Bottle Color
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
DWK218012417
Duran
®
Laboratory Bottle,
neck Joints: threaded (GL45), clear bottle, graduated, with screw cap to drain contents, with high temperature red PBT screw-cap and ETFE pouring ring, capacity 100 mL
Expand
View Pricing
DWK218015414
Duran
®
Laboratory Bottle,
clear, graduated, GL45, 1000mL, PBT screw cap, ETFE pouring ring, 10EA
Expand
View Pricing
DWK218019251
Duran
®
Laboratory Bottle,
capacity 25000 mL, clear bottle, graduated, neck Joints: threaded (GL45), with screw cap to drain contents, WITH SCREW-CAP AND POURING RING FROM PP (BLUE)
Expand
View Pricing
DWK218017312
Duran
®
Laboratory Bottle,
neck Joints: threaded (GL45), clear bottle, graduated, with screw cap to drain contents, WITH HIGH TEMPERATURE RED PBT SCREW-CAP AND ETFE POURING RING, capacity 5000 mL
Expand
View Pricing
DWK218014418
Duran
®
Laboratory Bottle,
neck Joints: threaded (GL45), clear bottle, graduated, with screw cap to drain contents, WITH HIGH TEMPERATURE RED PBT SCREW-CAP AND ETFE POURING RING, capacity 500 mL
Expand
View Pricing
DWK218013619
Duran
®
Laboratory Bottle,
neck Joints: threaded (GL45), clear bottle, graduated, with screw cap to drain contents, WITH HIGH TEMPERATURE RED PBT SCREW-CAP AND ETFE POURING RING, capacity 250 mL
Expand
View Pricing
DWK218016316
Duran
®
Laboratory Bottle,
clear bottle, graduated, neck Joints: threaded (GL45), with screw cap to drain contents, WITH HIGH TEMPERATURE RED PBT SCREW-CAP AND ETFE POURING RING, capacity 2000 mL
Expand
View Pricing
DWK218018617
Duran
®
Laboratory Bottle,
neck Joints: threaded (GL45), clear bottle, graduated, with screw cap to drain contents, WITH HIGH TEMPERATURE RED PBT SCREW-CAP AND ETFE POURING RING, capacity 10000 mL
Expand
View Pricing