
Doxycycline monohydrate

Synonym(s):
Doxycycline monohydrate
Empirical Formula (Hill Notation):
C22H24N2O8 · H2O
CAS Number:
Molecular Weight:
462.45
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Activity Spectrum
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing