Skip to Content
Merck
Doxazosin Related Compound D certified reference material, pharmaceutical secondary standard

Doxazosin Related Compound D

Doxazosin Related Compound D certified reference material, pharmaceutical secondary standard
Synonym(s):
1,4-Benzodioxan-2-carboxylic acid
Empirical Formula (Hill Notation):
C9H8O4
CAS Number:
Molecular Weight:
180.16

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2