
DMT-2′O-TBDMS-rA(bz) Phosphoramidite

Synonym(s):
DMT-2’O-TBDMS-rA(bz) Amidite, N-benzoyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-[(1,1-dimethylethyl)dimethylsilyl]-adenosine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]
Empirical Formula (Hill Notation):
C53H66N7O8PSi
CAS Number:
Molecular Weight:
988.19
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Quality Segment
Technique
Showing 1-4 of 4