
DMT-2′O-Methyl-rA(bz) Phosphoramidite

Synonym(s):
DMT-2′-O-Me-rA(bz) amidite, N-benzoyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-methyl-adenosine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]
Empirical Formula (Hill Notation):
C48H54N7O8P
CAS Number:
Molecular Weight:
887.96
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Quality Segment
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing