Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
(±)-Verapamil hydrochloride
Synonym(s):
5-[
N
-(3,4-Dimethoxyphenylethyl)methylamino]-2-(3,4-dimethoxyphenyl)-2-isopropylvaleronitrile hydrochloride
Linear Formula:
(CH
3
O)
2
C
6
H
3
CH
2
CH
2
N(CH
3
)(CH
2
)
3
C[C
6
H
3
(OCH
3
)
2
][CH(CH
3
)
2
]CN · HCl
CAS Number:
152-11-4
Molecular Weight:
491.06
EC Number:
205-800-5
Products
Product Category
bioactive small molecules (1)
Brand
Supelco (1)
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
certified reference material (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
V4629
(±)-Verapamil hydrochloride,
≥99% (titration), powder, α1-adrenoceptor antagonist
Expand
View Pricing
PHR1131
(±)-Verapamil hydrochloride,
Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing