Skip to Content
Merck
Discovery® C8 Supelguard Guard Cartridge 5 μm particle size, L × I.D. 2 cm × 4 mm

Discovery® C8 (5 µm) HPLC Columns

Discovery® C8 Supelguard Guard Cartridge 5 μm particle size, L × I.D. 2 cm × 4 mm

Products

Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Application
Column Material
Package Size
Particle Size (µm)
Pore Size (Å)
Technique