Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Discharge tube with recirculation valve for Dispensette
®
S
Products
Product Category
dispensers (10)
Manufacturer
Length (mm)
Package Size
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
BR708119
Discharge tube with recirculation valve for Dispensette
®
S,
Organic, 25, 50 and 100 mL, standard, L 135 mm
Expand
View Pricing
BR708116
Discharge tube with recirculation valve for Dispensette
®
S,
Organic, 25, 50 and 100 mL,
fine tip
, L 135 mm
Expand
View Pricing
BR708104
Discharge tube with recirculation valve for Dispensette
®
S,
5 and 10 mL, standard, L 105 mm
Expand
View Pricing
BR708114
Discharge tube with recirculation valve for Dispensette
®
S,
Organic, 5 and 10 mL, standard, L 105 mm
Expand
View Pricing
BR708102
Discharge tube with recirculation valve for Dispensette
®
S,
1,2,5 and 10 mL, fine tip, L 105 mm
Expand
View Pricing
BR708106
Discharge tube with recirculation valve for Dispensette
®
S,
25, 50 and 100 mL,
fine tip
, L 135 mm
Expand
View Pricing
BR708109
Discharge tube with recirculation valve for Dispensette
®
S,
25, 50 and 100 mL, standard, L 135 mm
Expand
View Pricing
BR708124
Discharge tube with recirculation valve for Dispensette
®
S,
Trace Analysis, 10 mL, PT/IR spring (Tantalum Spring), L 90 mm
Expand
View Pricing
BR708112
Discharge tube with recirculation valve for Dispensette
®
S,
Organic, capacity 5, 10 mL,
fine tip
, L 105 mm
Expand
View Pricing
BR708122
Discharge tube with recirculation valve for Dispensette
®
S,
Trace Analysis, 10 mL, PT/IR spring, L 90 mm
Expand
View Pricing