Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Direct Thermal WetGrip labels
Products
Product Category
labels (5)
Color
Manufacturer
Length (in)
Package Size
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
Z743504
Direct Thermal WetGrip labels,
Tag and spot, white, L × W 1.5 in. × 0.5 in. , Tag, diam. 0.375 in. , Spot, roll of 1000
Expand
View Pricing
Z743507
Direct Thermal WetGrip labels,
roll of 1000
Expand
View Pricing
Z743505
Direct Thermal WetGrip labels,
white, L × W 1.05 in. × 0.50 in., roll of 1000
Expand
View Pricing
Z743506
Direct Thermal WetGrip labels,
white, L × W 1.50 in. × 0.50 in., roll of 750
Expand
View Pricing
Z743509
Direct Thermal WetGrip labels,
white, diam. 0.375 in., roll of 1000
Expand
View Pricing