Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Direct thermal Cryo-Tags
®
and Cryo-Spot combo
Products
Product Category
labels (2)
Color
Manufacturer
Length (in)
Color
Package Size
Technique
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
Z743353
Direct thermal Cryo-Tags
®
and Cryo-Spot combo,
blue, L × W 1.50 in. × 0.50 in. × 3/8 in., spot combo, pkg of 1000 ea per roll (500 tag and spot sets)
Expand
View Pricing
Z743346
Direct thermal Cryo-Tags
®
and Cryo-Spot combo,
Nanodrop Lite format, white, L × W 1.50 in. × 0.50 in. × 3/8 in., spot combo, pkg of 1000 ea per roll (500 tag and spot sets)
Expand
View Pricing
Z743354
Direct thermal Cryo-Tags
®
and Cryo-Spot combo,
green, L × W 1.50 in. × 0.50 in. × 3/8 in., spot combo, pkg of 1000 ea per roll (500 tag and spot sets)
Expand
View Pricing
Z743358
Direct thermal Cryo-Tags
®
and Cryo-Spot combo,
yellow, L × W 1.50 in. × 0.50 in. × 3/8 in., spot combo, pkg of 1000 ea per roll (500 tag and spot sets)
Expand
View Pricing
Z743355
Direct thermal Cryo-Tags
®
and Cryo-Spot combo,
orange, L × W 1.50 in. × 0.50 in. × 3/8 in., spot combo, pkg of 1000 ea per roll (500 tag and spot sets)
Expand
View Pricing
Z743357
Direct thermal Cryo-Tags
®
and Cryo-Spot combo,
red, L × W 1.50 in. × 0.50 in. × 3/8 in., spot combo, pkg of 1000 ea per roll (500 tag and spot sets)
Expand
View Pricing
Z744110
Direct thermal Cryo-Tags
®
and Cryo-Spot combo,
white, W × L 0.94 in. × 0.50 in. × 3/8 in., spot combo, pack of 1000 ea (500 tag and spot sets)
Expand
View Pricing
Z744111
Direct thermal Cryo-Tags
®
and Cryo-Spot combo,
white, W × L 1.28 in. × 0.50 in. × 3/8 in., spot combo, pack of 1000 ea (500 tag and spot sets)
Expand
View Pricing