Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
DI-PAK
®
Purification Pack
Products
Product Category
polishers (2)
water pretreatment attachments (1)
Brand
Milli-Q (3)
Diameter (mm)
Compatibility
Quality Segment
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
CPDI000L1
DI-PAK
®
Purification Pack,
Large: 4.7 kg, Produces deionized water, for use with Milli-DI
®
systems
Expand
View Pricing
CPDI0NJM1
DI-PAK
®
Purification Pack,
Medium: 2.4 kg, Produces deionized water, for use with Milli-DI
®
systems
Expand
View Pricing
CPDI000S1
DI-PAK
®
Purification Pack,
Small: L - 12.7 cm, For the production of de-ionized water
Expand
View Pricing