
Dioctyl phthalate-3,4,5,6-d4

Linear Formula:
C6D4[CO2CH2(CH2)6CH3]2
CAS Number:
Molecular Weight:
394.58
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Format
Quality Segment
Isotope
Application
Markush Class
Markush Group
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing