Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dihydrostreptomycin sesquisulfate
Linear Formula:
C
21
H
41
N
7
O
12
· 3/2H
2
SO
4
CAS Number:
5490-27-7
Molecular Weight:
730.71
EC Number:
226-823-7
Products
Product Category
antibiotics (2)
Brand
Sigma-Aldrich (2)
Biological Source
Color
Formula Weight
Physical Form
Product Line
Purity
Activity Spectrum
Mode of Action
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
D7253
Dihydrostreptomycin sesquisulfate,
Expand
View Pricing
D5155
Dihydrostreptomycin sesquisulfate,
powder, suitable for cell culture, BioReagent
Expand
View Pricing