
Dihydroartemisinin

Synonym(s):
β-Dihydroartemisinin, (3R,5aS,6R,8aS,9R,12R,12aR)-Decahydro-10-hydroxy-3,6,9-trimethyl-3,12-epoxy-12H-pyrano[4,3-j]-1,2-benzodioxepin
Empirical Formula (Hill Notation):
C15H24O5
CAS Number:
Molecular Weight:
284.35
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing