Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Diethylamine
Synonym(s):
Diethylamine
Linear Formula:
(C
2
H
5
)
2
NH
CAS Number:
109-89-7
Molecular Weight:
73.14
EC Number:
203-716-3
Products
Product Category
organic bases (1)
Brand
Sigma-Aldrich (4)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Puriss (1)
Puriss P.A. (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
471216
Diethylamine,
≥99.5%
Expand
View Pricing
8.03010
Diethylamine,
for synthesis
Expand
View Pricing
31730
Diethylamine,
puriss. p.a., ≥99.5% (GC)
Expand
View Pricing
386456
Diethylamine,
purified by redistillation, 99.5%
Expand
View Pricing