Skip to Content
Merck
Di(2-pyridyl) ketone 99%

Di(2-pyridyl) ketone

Di(2-pyridyl) ketone 99%
Synonym(s):
2,2′-Carbonyldipyridine, 2,2′-Dipyridyl ketone
Empirical Formula (Hill Notation):
C11H8N2O
CAS Number:
Molecular Weight:
184.19
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing