Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Deuterium oxide
Synonym(s):
Heavy water, Water-d
2
, Deuterium oxide
Empirical Formula (Hill Notation):
D
2
O
CAS Number:
7789-20-0
Molecular Weight:
20.03
EC Number:
232-148-9
Products
Product Category
solvents (7)
stable isotopes (5)
Brand
Sigma-Aldrich (9)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Technique
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
151882
Deuterium oxide,
99.9 atom % D
Expand
View Pricing
450510
Deuterium oxide,
99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-
2,2,3,3-d
4
acid, sodium salt
Expand
View Pricing
1.13366
Deuterium oxide,
deuteration degree min. 99.9% for NMR spectroscopy MagniSolv
™
Expand
View Pricing
293040
Deuterium oxide,
99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-
2,2,3,3-d
4
acid, sodium salt
Expand
View Pricing
1.03428
Deuterium oxide,
deuteration degree min. 99.95% for NMR spectroscopy MagniSolv
™
Expand
View Pricing
756822
Deuterium oxide,
filtered, 99.8 atom % D
Expand
View Pricing
343773
Deuterium oxide,
99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Expand
View Pricing
613428
Deuterium oxide,
70 atom % D
Expand
View Pricing
613436
Deuterium oxide,
60 atom % D
Expand
View Pricing