Skip to Content
Merck
vibrant-m

Deferoxamine for peak identification CRS 1

vibrant-m
Synonym(s):
Desferrioxamine B, 30-Amino-3,14,25-trihydroxy-3,9,14,20,25-pentaazatriacontane-2,10,13,21,24-pentone, Deferoxamine B
Empirical Formula (Hill Notation):
C25H48N6O8
CAS Number:
Molecular Weight:
560.68

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1