Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
D-(+)-Trehalose dihydrate
Synonym(s):
α,α-Trehalose, α-D-Glucopyranosyl-α-D-glucopyranoside
Empirical Formula (Hill Notation):
C
12
H
22
O
11
· 2H
2
O
CAS Number:
6138-23-4
Molecular Weight:
378.33
EC Number:
202-739-6
Products
Product Category
carbohydrates (5)
disaccharides (3)
Brand
Millipore (1)
Sigma-Aldrich (4)
Biological Source
Color
Formula Weight
Physical Form
Product Line
Purity
Research Area
Tissue
Quality Segment
Application
Optical Isomer
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
T9531
D-(+)-Trehalose dihydrate,
≥99% (HPLC), from
Saccharomyces cerevisiae
Expand
View Pricing
T0167
D-(+)-Trehalose dihydrate,
from
Saccharomyces cerevisiae
, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Expand
View Pricing
T5251
D-(+)-Trehalose dihydrate,
≥98.5% (HPLC), from
Saccharomyces cerevisiae
Expand
View Pricing
T9449
D-(+)-Trehalose dihydrate,
≥99% (HPLC), from starch
Expand
View Pricing
90210
D-(+)-Trehalose dihydrate,
suitable for microbiology, ≥99.0%, composed of two α-glucose units that is used as a protectant, stabilizer and to support proper folding
Expand
View Pricing