Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
D-(+)-Galactosamine hydrochloride
Synonym(s):
2-Amino-2-deoxy-D-galactopyranose hydrochloride, D-Chondrosamine hydrochloride
Empirical Formula (Hill Notation):
C
6
H
13
NO
5
· HCl
CAS Number:
1772-03-8
Molecular Weight:
215.63
EC Number:
217-198-1
Products
Product Category
carbohydrates (1)
media supplements (1)
monosaccharides (1)
Brand
Sigma-Aldrich (2)
Biological Source
Color
Formula Weight
Physical Form
Product Line
Purity
Tissue
Optical Isomer
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
G0500
D-(+)-Galactosamine hydrochloride,
≥99% (HPLC)
Expand
View Pricing
G1639
D-(+)-Galactosamine hydrochloride,
BioReagent, suitable for cell culture
Expand
View Pricing