Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cytochrome P450 human
Products
Product Category
enzymes (4)
proteins (1)
Brand
Sigma-Aldrich (6)
Biological Source
Gene Alias
Physical Form
Recombinant Host
Quality Segment
Application
Package Size
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
C4982
Cytochrome P450 human,
3A4 isozyme microsomes, with P450 reductase and cytochrome b
5
, recombinant, expressed in baculovirus infected insect cells (BTI-TN-5B1-4)
Expand
View Pricing
C3735
Cytochrome P450 human,
1A1 Isozyme Microsomes, with P450 Reductase, recombinant, expressed in baculovirus infected insect cells (BTI-TN-5B1-4)
Expand
View Pricing
C5614
Cytochrome P450 human,
1A2 Isozyme Microsomes, with P450 Reductase and cytochrome b
5
, recombinant, expressed in baculovirus infected insect cells (BTI-TN-5B1-4)
Expand
View Pricing
C5740
Cytochrome P450 human,
2E1 isozyme microsomes, with P450 Reductase and cytochrome b
5
, recombinant, expressed in baculovirus infected insect cells (BTI-TN-5B1-4)
Expand
View Pricing
C5235
Cytochrome P450 human,
4F3B isozyme microsomes, with P450 Reductase and cytochrome b
5
, recombinant, expressed in baculovirus infected insect cells (BTI-TN-5B1-4)
Expand
View Pricing
C3860
Cytochrome P450 human,
1B1 Isozyme Microsomes, with P450 Reductase, recombinant, expressed in baculovirus infected insect cells (BTI-TN-5B1-4)
Expand
View Pricing