Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cyclohexylmagnesium chloride solution
Empirical Formula (Hill Notation):
C
6
H
11
ClMg
CAS Number:
931-51-1
Molecular Weight:
142.91
Products
Brand
Sigma-Aldrich (2)
Sustainability
Feature
Formula Weight
Physical Form
Reaction Type
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
774677
Cyclohexylmagnesium chloride solution,
1.0 M in 2-methyltetrahydrofuran
Expand
View Pricing
224413
Cyclohexylmagnesium chloride solution,
2.0 M in diethyl ether
Expand
View Pricing
752134
Cyclohexylmagnesium chloride solution,
1.3 M in THF/toluene (1:1)
Expand
View Pricing