Skip to Content
Products
Cart
0
AE
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Back
Cryo-gloves
®
Products
Product Category
gloves (8)
Brand
Sigma-Aldrich (5)
Manufacturer
Color
Size
Sterility
Technique
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
Z183512
Cryo-gloves
®
,
size M, water-resistant
Expand
View Pricing
Z183520
Cryo-gloves
®
,
size L, water-resistant
Expand
View Pricing
Z183571
Cryo-gloves
®
,
size XL, waterproof
Expand
View Pricing
Z183563
Cryo-gloves
®
,
size L, waterproof
Expand
View Pricing
Z183555
Cryo-gloves
®
,
size M, waterproof
Expand
View Pricing
Z183490
Cryo-gloves
®
,
size S, water-resistant
Expand
View Pricing
Z183539
Cryo-gloves
®
,
size XL, water-resistant
Expand
View Pricing
Z183547
Cryo-gloves
®
,
size S, waterproof
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
English - EN
Learn More