Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Crimp seals with PTFE/rubber septa
Products
Product Category
septa (2)
Brand
Supelco (2)
Diameter (mm)
Closure Type
Color
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
27454-U
Crimp seals with PTFE/rubber septa,
seal diam. × thickness 20 mm × 3.0 mm,
pressure release open center
, PTFE/rubber septum, pkg of 100 ea
Expand
View Pricing
33233-U
Crimp seals with PTFE/rubber septa,
silver aluminum seal, seal diam. × thickness 11 mm × 1.0 mm, opening size 4.8 mm
Expand
View Pricing